siRNA Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013365.2 | NP_001013383.1 | 3 | 841 | ||
| GenBank mRNA | AK087945.1 | - | 1 | 251 | ||
| AK155637.1 | - | 3 | 792 | |||
| BC099866.1 | - | 3 | 793 | 40040107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osm| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013365.2 | NP_001013383.1 | 3 | 841 | ||
| GenBank mRNA | AK087945.1 | - | 1 | 251 | ||
| AK155637.1 | - | 3 | 792 | |||
| BC099866.1 | - | 3 | 793 | 40040107 | ||