siRNA Details
| Gene Symbol: Foxred2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017983.2 | NP_001017983.2 | 9 | 4417 | ||
| NM_001168260.1 | NP_001161732.1 | 9 | 4515 | |||
| GenBank mRNA | AK089177.1 | - | 2 | 2806 | ||
| AK140027.1 | - | 9 | 4377 | |||
| AK171308.1 | - | 2 | 2806 | |||
| BC086662.1 | - | 9 | 4347 | 30549783 | ||

Foxred2
Genomic Map