siRNA Details
| Gene Symbol: Cxx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018063.1 | NP_001018073.1 | 1 | 821 | ||
| GenBank mRNA | AK003646.1 | - | 1 | 800 | ||
| AK206042.1 | - | 1 | 189 | |||
| BC029838.1 | - | 1 | 821 | 3967024 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxx1b| Gene Symbol: Cxx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018063.1 | NP_001018073.1 | 1 | 821 | ||
| GenBank mRNA | AK003646.1 | - | 1 | 800 | ||
| AK206042.1 | - | 1 | 189 | |||
| BC029838.1 | - | 1 | 821 | 3967024 | ||