siRNA Details
| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024560.2 | NP_001019731.1 | 12 | 1226 | ||
| GenBank mRNA | AK037118.1 | - | 1800 | |||
| AK047234.1 | - | 12 | 1202 | |||
| AK143441.1 | - | 10 | 1921 | |||
| BC048922.1 | - | 12 | 1226 | 6311864 | ||

Snx32
Genomic Map