siRNA Details
| Gene Symbol: Hoxc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024842.1 | NP_001020013.1 | 1 | 422 | ||
| GenBank mRNA | BC139233.1 | - | 1 | 468 | 8862255 | |
| BC139269.1 | - | 1 | 468 | 8862291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc11| Gene Symbol: Hoxc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024842.1 | NP_001020013.1 | 1 | 422 | ||
| GenBank mRNA | BC139233.1 | - | 1 | 468 | 8862255 | |
| BC139269.1 | - | 1 | 468 | 8862291 | ||