siRNA Details
| Gene Symbol: Cfhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025575.2 | NP_001020746.1 | 6 | 1455 | ||
| GenBank mRNA | AJ565908.1 | - | 6 | 1404 | ||
| AK149511.1 | - | 6 | 1446 | |||
| BC026782.1 | - | 1972 | ||||
| M29010.1 | - | 2130 | ||||

Cfhr2
Genomic Map