siRNA Details
| Gene Symbol: Gm527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025605.1 | NP_001020776.1 | 2 | 341 | ||
| GenBank mRNA | BC099503.1 | - | 2 | 341 | 30940052 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm527| Gene Symbol: Gm527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025605.1 | NP_001020776.1 | 2 | 341 | ||
| GenBank mRNA | BC099503.1 | - | 2 | 341 | 30940052 | |