siRNA Details
Gene Symbol: Gm5168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025607.3 | NP_001020778.1 | 2 | 160 | ||
GenBank mRNA | BC099542.1 | - | 2 | 160 | 30917040 |
Gene Symbol: Gm4297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100446.2 | NP_001093916.1 | 1 | 101 |
