siRNA Details
Gene Symbol: Gm5168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025607.3 | NP_001020778.1 | 9 | 785 | ||
GenBank mRNA | BC099542.1 | - | 9 | 785 | 30917040 |
Gene Symbol: Gm1993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102677.2 | NP_001096147.1 | 9 | 866 |
Gene Symbol: Gm5935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081657.1 | NP_001075126.1 | 9 | 835 | ||
GenBank mRNA | BC100411.1 | - | 9 | 835 | 30916114 |
