siRNA Details
Gene Symbol: Psg26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029893.1 | NP_001025064.1 | 5 | 1959 | ||
GenBank mRNA | BC099404.1 | - | 5 | 1959 | 30787301 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psg26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029893.1 | NP_001025064.1 | 5 | 1959 | ||
GenBank mRNA | BC099404.1 | - | 5 | 1959 | 30787301 |