siRNA Details
| Gene Symbol: Kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030292.1 | NP_001025463.1 | 2 | 736 | ||
| GenBank mRNA | BC147255.1 | - | 2 | 835 | 9056306 | |
| BC147256.1 | - | 2 | 835 | 9056288 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk15| Gene Symbol: Kcnk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030292.1 | NP_001025463.1 | 2 | 736 | ||
| GenBank mRNA | BC147255.1 | - | 2 | 835 | 9056306 | |
| BC147256.1 | - | 2 | 835 | 9056288 | ||