siRNA Details
| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308101.1 | NP_001295030.1 | 7 | 2410 | ||
| NM_020640.3 | NP_065691.2 | 7 | 2392 | |||
| XM_017006635.1 | XP_016862124.1 | 8 | 2922 | |||
| GenBank mRNA | AF292100.2 | - | 7 | 2387 | ||
| AK025764.1 | - | 5 | 1956 | |||

DCUN1D1
Genomic Map