siRNA Details
| Gene Symbol: Sh2d1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033499.1 | NP_001028671.1 | 3 | 271 | ||
| GenBank mRNA | DQ131181.1 | - | 3 | 271 | ||
| DQ140178.1 | - | 3 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d1b2| Gene Symbol: Sh2d1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033499.1 | NP_001028671.1 | 3 | 271 | ||
| GenBank mRNA | DQ131181.1 | - | 3 | 271 | ||
| DQ140178.1 | - | 3 | 271 | |||