siRNA Details
| Gene Symbol: Slc2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033633.3 | NP_001028805.2 | 2 | 791 | ||
| GenBank mRNA | AJ874111.1 | - | 1 | 111 | ||
| AK147341.1 | - | 2 | 792 | |||
| BC119586.1 | - | 3 | 358 | 40058865 | ||
| BC119587.1 | - | 2 | 300 | |||
| BC131681.1 | - | 2 | 195 | |||

Slc2a13
Genomic Map