siRNA Details
| Gene Symbol: Slc2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033633.3 | NP_001028805.2 | 7, 8 | 1584 | ||
| GenBank mRNA | AK147341.1 | - | 7, 8 | 1585 | ||
| BC119586.1 | - | 8, 9 | 1151 | 40058865 | ||
| BC119587.1 | - | 7, 8 | 1093 | |||

Slc2a13
Genomic Map