siRNA Details
| Gene Symbol: Fbxw10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033669.2 | NP_001028841.2 | 1 | 497 | ||
| NM_001291441.1 | NP_001278370.1 | 1 | 497 | |||
| GenBank mRNA | AK132680.1 | - | 1 | 498 | ||
| BC085108.1 | - | 1 | 495 | |||
| BC141212.1 | - | 1 | 488 | 9055824 | ||

Fbxw10
Genomic Map