siRNA Details
| Gene Symbol: Vrtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033776.2 | NP_001028948.1 | 2 | 323 | ||
| NM_001168588.1 | NP_001162060.1 | 3 | 735 | |||
| NM_001168589.2 | NP_001162061.1 | 2 | 393 | |||
| GenBank mRNA | BC103771.1 | - | 2 | 323 | 30935479 | |

Vrtn
Genomic Map