siRNA Details
| Gene Symbol: Ctrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033875.2 | NP_001029047.1 | 7, 8 | 888 | ||
| GenBank mRNA | AK007772.1 | - | 6, 7 | 716 | ||
| BC103715.1 | - | 7, 8 | 886 | 6430373 | ||
| BC115516.1 | - | 7, 8 | 888 | 40101127 | ||
| BC115517.1 | - | 7, 8 | 888 | 40101128 | ||

Ctrc
Genomic Map