siRNA Details
| Gene Symbol: 1700024G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034037.1 | NP_001029209.1 | 2 | 164 | ||
| GenBank mRNA | AK006301.1 | - | 2 | 183 | ||
| AK007014.1 | - | 2 | 182 | |||
| AK019799.1 | - | 2 | 171 | |||
| BC107222.2 | - | 2 | 161 | 40051877 | ||
| BC107223.1 | - | 2 | 164 | 40051878 | ||

1700024G13Rik
Genomic Map