siRNA Details
Gene Symbol: Gm13119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034101.2 | NP_001029273.2 | 4 | 1236 | ||
GenBank mRNA | BC100406.1 | - | 3 | 1236 | 30727972 |
Gene Symbol: Pramef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126315.2 | NP_001119787.1 | 3 | 1240 |
