siRNA Details
| Gene Symbol: Gm5941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034103.1 | NP_001029275.1 | 147 | |||
| GenBank mRNA | BC051629.1 | - | 92 | |||
| BC100438.1 | - | 147 | 30916603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5941| Gene Symbol: Gm5941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034103.1 | NP_001029275.1 | 147 | |||
| GenBank mRNA | BC051629.1 | - | 92 | |||
| BC100438.1 | - | 147 | 30916603 | |||