siRNA Details
| Gene Symbol: Zfp345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034900.3 | NP_001030072.2 | 4 | 2649 | ||
| GenBank mRNA | AK144819.1 | - | 4 | 2649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp345| Gene Symbol: Zfp345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034900.3 | NP_001030072.2 | 4 | 2649 | ||
| GenBank mRNA | AK144819.1 | - | 4 | 2649 | ||