siRNA Details
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007749.3 | NP_031775.1 | 2 | 253 | ||
GenBank mRNA | AK002494.1 | - | 2 | 220 | ||
AK027989.1 | - | 2 | 215 | |||
AK131871.1 | - | 2 | 249 | |||
AK153444.1 | - | 2 | 225 | |||
AK168077.1 | - | 2 | 241 | |||
AV056265.1 | - | 2 | 93 | |||
BB696274.1 | - | 2 | 263 | |||
BC010772.1 | - | 2 | 235 | 4216583 | ||
BC086792.1 | - | 2 | 225 | 6411273 | ||
X52940.1 | - | 2 | 219 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
