siRNA Details
Gene Symbol: Ifna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008334.3 | NP_032360.2 | 1 | 561 | ||
GenBank mRNA | BC130231.1 | - | 1 | 562 | 40141930 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008334.3 | NP_032360.2 | 1 | 561 | ||
GenBank mRNA | BC130231.1 | - | 1 | 562 | 40141930 |