siRNA Details
| Gene Symbol: Mt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008630.2 | NP_032656.1 | 2 | 297 | ||
| GenBank mRNA | AK002567.1 | - | 2 | 132 | ||
| BC031758.1 | - | 2 | 114 | 2651471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mt2| Gene Symbol: Mt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008630.2 | NP_032656.1 | 2 | 297 | ||
| GenBank mRNA | AK002567.1 | - | 2 | 132 | ||
| BC031758.1 | - | 2 | 114 | 2651471 | ||