siRNA Details
| Gene Symbol: TTYH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005367.2 | NP_001005367.1 | 13 | 1846 | ||
| NM_001201461.1 | NP_001188390.1 | 14 | 1895 | |||
| NM_020659.3 | NP_065710.1 | 14 | 1892 | |||
| XM_017027025.1 | XP_016882514.1 | 13 | 1690 | |||
| XM_017027026.1 | XP_016882515.1 | 14 | 1736 | |||
| GenBank mRNA | AB188496.1 | - | 14 | 1895 | ||
| AF177909.1 | - | 14 | 1805 | |||
| BC019358.1 | - | 11 | 1228 | 3960435 | ||

TTYH1
Genomic Map