siRNA Details
| Gene Symbol: Tlx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009392.2 | NP_033418.1 | 3 | 1305 | ||
| GenBank mRNA | AK013072.1 | - | 2 | 1381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlx2| Gene Symbol: Tlx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009392.2 | NP_033418.1 | 3 | 1305 | ||
| GenBank mRNA | AK013072.1 | - | 2 | 1381 | ||