siRNA Details
| Gene Symbol: Tgoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009444.1 | NP_033470.1 | 2 | 181 | ||
| GenBank mRNA | AK145212.1 | - | 2 | 168 | ||
| D50032.1 | - | 2 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgoln2| Gene Symbol: Tgoln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009444.1 | NP_033470.1 | 2 | 181 | ||
| GenBank mRNA | AK145212.1 | - | 2 | 168 | ||
| D50032.1 | - | 2 | 181 | |||