siRNA Details
| Gene Symbol: Gm4836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009529.3 | NP_033555.1 | 9 | 866 | ||
| GenBank mRNA | BC061169.1 | - | 9 | 866 | 6774630 | |
| Gene Symbol: Gm10487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100609.1 | NP_001094079.1 | 9 | 866 | ||

Gm4836
Genomic Map