siRNA Details
| Gene Symbol: Zfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044697.2 | NP_001038162.1 | 6 | 2616 | ||
| NM_001044698.2 | NP_001038163.1 | 6 | 2612 | |||
| NM_001044700.2 | NP_001038165.1 | 6 | 2568 | |||
| NM_001294323.1 | NP_001281252.1 | 6 | 2575 | |||
| NM_178447.3 | NP_848542.1 | 7 | 2653 | |||
| GenBank mRNA | AK036776.1 | - | 7 | 2644 | ||
| AK131978.1 | - | 6 | 2564 | |||
| AK141471.1 | - | 6 | 2592 | |||
| BC052029.1 | - | 4 | 2316 | |||
| BC057171.1 | - | 6 | 2582 | |||
| BC089506.1 | - | 6 | 2547 | 30247022 | ||
| Y00850.1 | - | 5 | 2110 | |||

Zfp2
Genomic Map