siRNA Details
| Gene Symbol: Arsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009712.3 | NP_033842.3 | 8 | 1859 | ||
| GenBank mRNA | AK140163.1 | - | 8 | 1914 | ||
| AK154098.1 | - | 8 | 1779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arsb| Gene Symbol: Arsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009712.3 | NP_033842.3 | 8 | 1859 | ||
| GenBank mRNA | AK140163.1 | - | 8 | 1914 | ||
| AK154098.1 | - | 8 | 1779 | |||