siRNA Details
| Gene Symbol: Cdk5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009872.3 | NP_034002.2 | 1 | 782 | ||
| GenBank mRNA | BC100714.1 | - | 2 | 302 | ||
| BC100715.1 | - | 2 | 461 | |||
| BC103550.1 | - | 2 | 444 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109309.

Cdk5r2
Genomic Map