siRNA Details
| Gene Symbol: Cdk5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009872.3 | NP_034002.2 | 1 | 1155 | ||
| GenBank mRNA | BC100712.1 | - | 2 | 620 | ||
| BC100714.1 | - | 2 | 675 | |||
| BC100715.1 | - | 2 | 834 | |||
| BC103550.1 | - | 2 | 817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk5r2| Gene Symbol: Cdk5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009872.3 | NP_034002.2 | 1 | 1155 | ||
| GenBank mRNA | BC100712.1 | - | 2 | 620 | ||
| BC100714.1 | - | 2 | 675 | |||
| BC100715.1 | - | 2 | 834 | |||
| BC103550.1 | - | 2 | 817 | |||