siRNA Details
| Gene Symbol: Cyp2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009997.2 | NP_034127.2 | 2 | 243 | ||
| GenBank mRNA | BC010761.1 | - | 2 | 234 | 4239813 | |
| BC063778.1 | - | 2 | 266 | 5044482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2a4| Gene Symbol: Cyp2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009997.2 | NP_034127.2 | 2 | 243 | ||
| GenBank mRNA | BC010761.1 | - | 2 | 234 | 4239813 | |
| BC063778.1 | - | 2 | 266 | 5044482 | ||