siRNA Details
| Gene Symbol: Usp17lc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U70369.1 | - | 55 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp17lc| Gene Symbol: Usp17lc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U70369.1 | - | 55 | |||