siRNA Details
| Gene Symbol: Klk1b9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010116.1 | NP_034246.1 | 824 | |||
| GenBank mRNA | BC024624.1 | - | 803 | 4914236 | ||
| BC048869.1 | - | 788 | 4164261 | |||
| M17962.1 | - | 824 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b9| Gene Symbol: Klk1b9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010116.1 | NP_034246.1 | 824 | |||
| GenBank mRNA | BC024624.1 | - | 803 | 4914236 | ||
| BC048869.1 | - | 788 | 4164261 | |||
| M17962.1 | - | 824 | ||||