siRNA Details
| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271962.1 | NP_001258891.1 | 10 | 988 | ||
| NM_001271963.1 | NP_001258892.1 | 10 | 1164 | |||
| NM_010165.3 | NP_034295.1 | 10 | 1165 | |||
| GenBank mRNA | AK137718.1 | - | 10 | 1160 | ||
| AK141765.1 | - | 10 | 1164 | |||
| AY212511.1 | - | 7 | 679 | |||
| BC003755.1 | - | 10 | 1165 | 3497245 | ||
| U61111.1 | - | 9 | 906 | |||
| U71208.1 | - | 10 | 990 | |||
| U81603.1 | - | 9 | 905 | |||

Eya2
Genomic Map