siRNA Details
| Gene Symbol: Fcnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010190.1 | NP_034320.1 | 8 | 692 | ||
| GenBank mRNA | AF063217.1 | - | 8 | 665 | ||
| AK010913.1 | - | 7 | 691 | |||
| AK151676.1 | - | 8 | 700 | |||
| BC107220.1 | - | 8 | 692 | 40051808 | ||
| BC107221.1 | - | 8 | 692 | 40051810 | ||

Fcnb
Genomic Map