siRNA Details
| Gene Symbol: H2-T9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010399.2 | NP_034529.1 | 879 | |||
| GenBank mRNA | BC044208.1 | - | 890 | 5036676 | ||
| M35248.1 | - | 879 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-T9| Gene Symbol: H2-T9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010399.2 | NP_034529.1 | 879 | |||
| GenBank mRNA | BC044208.1 | - | 890 | 5036676 | ||
| M35248.1 | - | 879 | ||||