siRNA Details
| Gene Symbol: Irx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010573.2 | NP_034703.2 | 2 | 804 | ||
| GenBank mRNA | AF165984.1 | - | 2 | 569 | ||
| AK133937.1 | - | 2 | 804 | |||
| Y15002.1 | - | 1 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irx1| Gene Symbol: Irx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010573.2 | NP_034703.2 | 2 | 804 | ||
| GenBank mRNA | AF165984.1 | - | 2 | 569 | ||
| AK133937.1 | - | 2 | 804 | |||
| Y15002.1 | - | 1 | 197 | |||