siRNA Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010584.3 | NP_034714.1 | 8 | 981 | ||
| GenBank mRNA | AB016496.1 | - | 8 | 980 | ||
| AK007447.1 | - | 8 | 981 | |||
| AY157363.1 | - | 8 | 970 | |||
| AY157364.1 | - | 8 | 965 | |||
| AY619693.1 | - | 8 | 964 | |||
| BC150795.1 | - | 8 | 923 | 9087706 | ||

Itln1
Genomic Map