siRNA Details
| Gene Symbol: SPC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020675.3 | NP_065726.1 | 4 | 460 | ||
| XM_011511516.2 | XP_011509818.1 | 4 | 469 | |||
| GenBank mRNA | AF225416.1 | - | 4 | 442 | ||
| AK291093.1 | - | 4 | 471 | |||
| AY456388.1 | - | 3 | 329 | |||
| BC022255.1 | - | 4 | 460 | 4738187 | ||
| CR457197.1 | - | 3 | 329 | |||

SPC25
Genomic Map