siRNA Details
Gene Symbol: Myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010860.3 | NP_034990.1 | 5 | 388 | ||
GenBank mRNA | AK143885.1 | - | 5 | 418 | ||
AK159044.1 | - | 1 | 1790 | |||
AK167106.1 | - | 5 | 418 | |||
AK167191.1 | - | 5 | 424 | |||
AK168450.1 | - | 5 | 439 | |||
AK168869.1 | - | 5 | 420 | |||
BC026760.1 | - | 5 | 388 | 3990090 | ||
BC081470.1 | - | 5 | 405 | 6824167 | ||
BC089485.1 | - | 5 | 411 | 30277988 | ||
BC092214.1 | - | 5 | 396 | 5684607 | ||
U04443.1 | - | 3 | 344 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
