siRNA Details
| Gene Symbol: Olfr242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010974.1 | NP_035104.1 | 764 | |||
| GenBank mRNA | X89693.1 | - | 390 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr242| Gene Symbol: Olfr242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010974.1 | NP_035104.1 | 764 | |||
| GenBank mRNA | X89693.1 | - | 390 | |||