siRNA Details
Gene Symbol: Sprr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011469.3 | NP_035599.2 | 601 | |||
GenBank mRNA | AY158987.1 | - | 601 | |||
BC132518.1 | - | 587 | 40130795 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sprr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011469.3 | NP_035599.2 | 601 | |||
GenBank mRNA | AY158987.1 | - | 601 | |||
BC132518.1 | - | 587 | 40130795 |