siRNA Details
| Gene Symbol: Xlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291747.1 | NP_001278676.1 | 2 | 106 | ||
| NM_001291748.1 | NP_001278677.1 | 2 | 114 | |||
| NM_011725.4 | NP_035855.1 | 2 | 154 | |||
| GenBank mRNA | AA623498.1 | - | 2 | 68 | ||
| AK012549.1 | - | 2 | 145 | |||
| AK035025.1 | - | 2 | 116 | |||
| AK035341.1 | - | 2 | 123 | |||
| AK088930.1 | - | 2 | 58 | |||
| BC005560.1 | - | 2 | 141 | 3965533 | ||
| X07967.1 | - | 2 | 89 | |||
| Gene Symbol: 3830403N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014417.1 | - | 1 | 134 | ||

Xlr
Genomic Map