siRNA Details
| Gene Symbol: Xlr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110784.1 | NP_001104254.1 | 5, 6 | 453 | ||
| GenBank mRNA | BC003812.1 | - | 5, 6 | 399 | 3601144 | |
| BC093495.1 | - | 376 | 3593889 | |||
| BC107255.1 | - | 362 | 40052405 | |||
| BC107283.1 | - | 363 | 40052875 | |||
| BC127145.1 | - | 363 | 40052876 | |||
| BC127146.1 | - | 364 | 40052877 | |||
| BQ885410.1 | - | 5, 6 | 393 | |||
| DV662513.1 | - | 5, 6 | 401 | |||
| U02599.1 | - | 4, 5 | 346 | |||

Xlr3a
Genomic Map