siRNA Details
| Gene Symbol: Zfp97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011765.5 | NP_035895.3 | 4 | 856 | ||
| GenBank mRNA | L20450.1 | - | 824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp97| Gene Symbol: Zfp97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011765.5 | NP_035895.3 | 4 | 856 | ||
| GenBank mRNA | L20450.1 | - | 824 | |||