siRNA Details
| Gene Symbol: Rtn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025364.3 | NP_001020535.1 | 2 | 124 | ||
| NM_013648.6 | NP_038676.1 | 5 | 1038 | |||
| GenBank mRNA | AF038538.1 | - | 5 | 994 | ||
| AF038539.1 | - | 2 | 63 | |||
| AK144271.1 | - | 5 | 1001 | |||
| AK160500.1 | - | 2 | 85 | |||
| BC031370.1 | - | 5 | 995 | 5359358 | ||
| W65979.1 | - | 2 | 124 | |||

Rtn2
Genomic Map