siRNA Details
| Gene Symbol: Gbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015739.2 | NP_056554.1 | 2 | 1237 | ||
| GenBank mRNA | AF547996.1 | - | 2 | 1237 | ||
| AY319256.1 | - | 2 | 1591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbx1| Gene Symbol: Gbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015739.2 | NP_056554.1 | 2 | 1237 | ||
| GenBank mRNA | AF547996.1 | - | 2 | 1237 | ||
| AY319256.1 | - | 2 | 1591 | |||